missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(Butylamino)ethanol 98.0+%, TCI America™
$39.14 - $111.84
Chemical Identifiers
| CAS | 111-75-1 |
|---|---|
| Molecular Formula | C6H15NO |
| Molecular Weight (g/mol) | 117.19 |
| MDL Number | MFCD00020570 |
| InChI Key | LJDSTRZHPWMDPG-UHFFFAOYSA-N |
| Synonym | N-Butylethanolamine |
| PubChem CID | 8132 |
| IUPAC Name | 2-(butylamino)ethan-1-ol |
| SMILES | CCCCNCCO |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
B102325ML
|
TCI America
B102325ML |
25 mL |
Each for $39.14
|
|
|||||
|
B1023500ML
|
TCI America
B1023500ML |
500 mL |
Each for $111.84
|
|
|||||
Chemical Identifiers
| 111-75-1 | |
| 117.19 | |
| LJDSTRZHPWMDPG-UHFFFAOYSA-N | |
| 8132 | |
| CCCCNCCO |
| C6H15NO | |
| MFCD00020570 | |
| N-Butylethanolamine | |
| 2-(butylamino)ethan-1-ol |
Specifications
| 111-75-1 | |
| 199°C | |
| MFCD00020570 | |
| 2735 | |
| LJDSTRZHPWMDPG-UHFFFAOYSA-N | |
| 2-(butylamino)ethan-1-ol | |
| 8132 | |
| ≥98.0% (GC,T) | |
| 2-(Butylamino)ethanol |
| Colorless | |
| C6H15NO | |
| 25 mL | |
| N-Butylethanolamine | |
| CCCCNCCO | |
| 117.19 | |
| 117.19 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0295
RTECSNumber : KK0175000
TSCA : Yes