missing translation for 'onlineSavingsMsg'
Learn More

2-(Butylamino)ethanol 98.0+%, TCI America™

Supplier:  TCI America B1023500ML

 View more versions of this product

Catalog No. B1023500ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2-(Butylamino)ethanol
Colorless
C6H15NO
500 mL
N-Butylethanolamine
CCCCNCCO
117.19
117.19
Liquid
111-75-1
199°C
MFCD00020570
2735
LJDSTRZHPWMDPG-UHFFFAOYSA-N
2-(butylamino)ethan-1-ol
8132
≥98.0% (GC,T)

Chemical Identifiers

111-75-1
117.19
LJDSTRZHPWMDPG-UHFFFAOYSA-N
8132
CCCCNCCO
C6H15NO
MFCD00020570
N-Butylethanolamine
2-(butylamino)ethan-1-ol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0295

RTECSNumber : KK0175000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions