missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Butanol, Analytical Standard, ≥99.8% (GC), MilliporeSigma™ Supelco™
Supplier: Merck Emd Millipore 9687010MLF
Specifications
| 78-92-2 | |
| 2.6 (vs. air) | |
| Neat | |
| MFCD00004569 | |
| 98°C | |
| 10 mL | |
| ≥99.8% (GC) |
| 12.5 mmHg (20°C) | |
| 0.808 g/mL (at 25°C) | |
| CH3CH2CH(OH)CH3 | |
| -115°C | |
| sec-Butyl alcohol | |
| 74.12 | |
| Analytical Standard |