missing translation for 'onlineSavingsMsg'
Learn More

2-Butanol, Analytical Standard, ≥99.8% (GC), MilliporeSigma™ Supelco™

Catalog No. p-7193501
Change view
Click to view available options
Quantity:
1 mL
10 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020408 1 mL
111018353 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111020408 Supplier Merck Emd Millipore Supplier No. 968701MLF
Only null left

Specifications

CAS 78-92-2
Vapor Pressure 12.5 mmHg (20°C)
Vapor Density 2.6 (vs. air)
Density 0.808 g/mL (at 25°C)
Physical Form Neat
Linear Formula CH3CH2CH(OH)CH3
MDL Number MFCD00004569
Melting Point -115°C
Boiling Point 98°C
Synonym sec-Butyl alcohol
Quantity 1 mL
Formula Weight 74.12
Percent Purity ≥99.8% (GC)
Grade Analytical Standard
Show More Show Less