missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromophenylhydrazine Hydrochloride Hydrate 98.0+%, TCI America™
Supplier: TCI America B214725G
Specifications
| 2-Bromophenylhydrazine Hydrochloride Hydrate | |
| 185°C | |
| MFCD00012926 | |
| 2-bromophenylhydrazine hydrochloride, 2-bromophenyl hydrazine hydrochloride, 2-bromophenylhydrazine hcl, hydrazine, 2-bromophenyl-, monohydrochloride, 1-2-bromophenyl hydrazine hydrochloride, 1-bromo-2-hydrazino benzene hydrochloride, pubchem4463, acmc-1b1zz, ksc270g5h, 2-bromophenylhydrazine, chloride | |
| C1=CC=C(C(=C1)NN)Br.Cl | |
| 223.498 | |
| 223.50 | |
| Crystalline Powder |
| 50709-33-6 | |
| C6H8BrClN2 | |
| 25 g | |
| PHCYUJRYSFMJMG-UHFFFAOYSA-N | |
| (2-bromophenyl)hydrazine;hydrochloride | |
| 2723912 | |
| ≥98.0% (HPLC,T) |
Chemical Identifiers
| 50709-33-6 | |
| 223.498 | |
| PHCYUJRYSFMJMG-UHFFFAOYSA-N | |
| 2723912 | |
| C1=CC=C(C(=C1)NN)Br.Cl |
| C6H8BrClN2 | |
| MFCD00012926 | |
| 2-bromophenylhydrazine hydrochloride, 2-bromophenyl hydrazine hydrochloride, 2-bromophenylhydrazine hcl, hydrazine, 2-bromophenyl-, monohydrochloride, 1-2-bromophenyl hydrazine hydrochloride, 1-bromo-2-hydrazino benzene hydrochloride, pubchem4463, acmc-1b1zz, ksc270g5h, 2-bromophenylhydrazine, chloride | |
| (2-bromophenyl)hydrazine;hydrochloride |
Safety and Handling
TSCA : No