missing translation for 'onlineSavingsMsg'
Learn More

2-Bromophenylhydrazine Hydrochloride Hydrate 98.0+%, TCI America™

Catalog No. p-7119511
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B21475G 5 g
B214725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B21475G Supplier TCI America Supplier No. B21475G

Chemical Identifiers

CAS 50709-33-6
Molecular Formula C6H8BrClN2
Molecular Weight (g/mol) 223.498
MDL Number MFCD00012926
InChI Key PHCYUJRYSFMJMG-UHFFFAOYSA-N
Synonym 2-bromophenylhydrazine hydrochloride, 2-bromophenyl hydrazine hydrochloride, 2-bromophenylhydrazine hcl, hydrazine, 2-bromophenyl-, monohydrochloride, 1-2-bromophenyl hydrazine hydrochloride, 1-bromo-2-hydrazino benzene hydrochloride, pubchem4463, acmc-1b1zz, ksc270g5h, 2-bromophenylhydrazine, chloride
PubChem CID 2723912
IUPAC Name (2-bromophenyl)hydrazine;hydrochloride
SMILES C1=CC=C(C(=C1)NN)Br.Cl

Specifications

Melting Point 185°C
Quantity 5 g
Formula Weight 223.50
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromophenylhydrazine Hydrochloride Hydrate