missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromophenol 98.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 194°C |
| Quantity | 100 g |
| UN Number | 2929 |
| Formula Weight | 173.01 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Liquid |
| Chemical Name or Material | 2-Bromophenol |
Safety and Handling
| EINECSNumber | (3)-0896 |
| RTECSNumber | SJ7875000 |
| TSCA | Yes |