missing translation for 'onlineSavingsMsg'
Learn More

2-Bromophenol 98.0+%, TCI America™

Catalog No. B063025G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B063025G 25 g
B0630100G 100 g
B0630500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B063025G Supplier TCI America Supplier No. B063025G

Chemical Identifiers

CAS 95-56-7
Molecular Formula C6H5BrO
Molecular Weight (g/mol) 173.009
MDL Number MFCD00002146
InChI Key VADKRMSMGWJZCF-UHFFFAOYSA-N
Synonym o-bromophenol, phenol, 2-bromo, bromophenol, 2-bromfenol, 2-bromo phenol, 2-bromo-phenol, phenol, o-bromo, o-bromo-phenol, phenol, bromo, 2-bromfenol czech
PubChem CID 7244
IUPAC Name 2-bromophenol
SMILES C1=CC=C(C(=C1)O)Br

Specifications

Color Yellow
Boiling Point 194°C
Quantity 25 g
UN Number 2929
Formula Weight 173.01
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Bromophenol
EINECSNumber (3)-0896
RTECSNumber SJ7875000
TSCA Yes