missing translation for 'onlineSavingsMsg'
Learn More

2-Bromononane, 97%

Catalog No. AAL0738706
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0738706 5 g
AAL0738703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0738706 Supplier Thermo Scientific Chemicals Supplier No. L0738706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2216-35-5 | C9H19Br | 207.155 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2216-35-5
Molecular Formula C9H19Br
Molecular Weight (g/mol) 207.155
MDL Number MFCD00017865
InChI Key JQEFZTLHNWFZDD-UHFFFAOYSA-N
Synonym sec-nonyl bromide, nonane, 2-bromo, acmc-1cs41
PubChem CID 98219
IUPAC Name 2-bromononane
SMILES CCCCCCCC(C)Br

Specifications

Density 1.081
Boiling Point 208°C to 209°C
Refractive Index 1.453
Quantity 5 g
Beilstein 1697559
Formula Weight 207.16
Percent Purity 97%
Chemical Name or Material 2-Bromononane
EINECSNumber 218-688-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only