accessibility menu, dialog, popup

UserName

H3CCH3Br

CAS RN 2216-35-5

IUPAC Name: 2-bromononane
Synonyms: 2-bromononane
Molecular Weight (g/mol): 207.16
Molecular Formula: C9H19Br
InChi Key: JQEFZTLHNWFZDD-UHFFFAOYNA-N
SMILES: CCCCCCCC(C)Br
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)