missing translation for 'onlineSavingsMsg'
Learn More

2-(Bromomethyl)benzeneboronic acid, 97%

Catalog No. AAH5246703
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5246703 1 g
AAH5246706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5246703 Supplier Thermo Scientific Chemicals Supplier No. H5246703

CAS: 91983-14-1 | C7H8BBrO2 | 214.85 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 91983-14-1
Molecular Formula C7H8BBrO2
Molecular Weight (g/mol) 214.85
MDL Number MFCD01318941
InChI Key MYVJCOQGXCONPE-UHFFFAOYSA-N
Synonym 2-bromomethylphenylboronic acid, 2-bromomethyl benzeneboronic acid, 2-bromomethyl phenylboronic acid, 2-bromomethyl phenyl boronic acid, 2-bromomethylphenylboronicacid, 2-boronobenzyl bromide, o-boronobenzyl bromide, 2-bromomethylphenyl boronic acid, pubchem7781
PubChem CID 2773278
IUPAC Name [2-(bromomethyl)phenyl]boronic acid
SMILES OB(O)C1=CC=CC=C1CBr

Specifications

Melting Point 145°C to 148°C
Quantity 1 g
Formula Weight 214.85
Percent Purity 97%
Chemical Name or Material 2-(Bromomethyl)benzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only