Learn More
2-Bromomethyl-1,3-dioxolane, 97%
CAS: 4360-63-8 | C4H7BrO2 | 167.002 g/mol
$129.59 - $1869.45
Chemical Identifiers
| CAS | 4360-63-8 |
|---|---|
| Molecular Formula | C4H7BrO2 |
| Molecular Weight (g/mol) | 167.002 |
| MDL Number | MFCD00003214 |
| InChI Key | CKIIJIDEWWXQEA-UHFFFAOYSA-N |
| Synonym | 2-bromomethyl-1,3-dioxolane, bromoacetaldehyde ethylene acetal, 1,3-dioxolane, 2-bromomethyl, 1,3-dioxolan-2-yl methyl bromide, bromomethyl dioxolane, acmc-1afrf, 2-bromomethyl-1,3-dioxolan, ksc495q7b, 2-bromomethyl-1,3 dioxolane, 2-bromomethyl-1, 3-dioxolane |
| PubChem CID | 78068 |
| IUPAC Name | 2-(bromomethyl)-1,3-dioxolane |
| SMILES | C1COC(O1)CBr |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1931214
|
Thermo Scientific Chemicals
A1931214 |
25 g |
Each for $129.59
|
|
|||||
|
AAA1931222
|
Thermo Scientific Chemicals
A1931222 |
100 g |
Each for $365.42
|
|
|||||
|
AAA1931236
|
Thermo Scientific Chemicals
A1931236 |
500 g |
Each for $1,869.45
|
|
|||||
Description
2-Bromomethyl-1,3-dioxolane is used in the preparation of alfa, beta-unsaturated aldehyde and 1, 4-two aldehyde monoacetals synthesis of monomers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Bromomethyl-1,3-dioxolane is used in the preparation of alfa, beta-unsaturated aldehyde and 1, 4-two aldehyde monoacetals synthesis of monomers.
Solubility
Immiscible with water.
Notes
Moisture sensitive. Incompatible with strong oxidizing agents, strong acids and strong bases.
Chemical Identifiers
| 4360-63-8 | |
| 167.002 | |
| CKIIJIDEWWXQEA-UHFFFAOYSA-N | |
| 78068 | |
| C1COC(O1)CBr |
| C4H7BrO2 | |
| MFCD00003214 | |
| 2-bromomethyl-1,3-dioxolane, bromoacetaldehyde ethylene acetal, 1,3-dioxolane, 2-bromomethyl, 1,3-dioxolan-2-yl methyl bromide, bromomethyl dioxolane, acmc-1afrf, 2-bromomethyl-1,3-dioxolan, ksc495q7b, 2-bromomethyl-1,3 dioxolane, 2-bromomethyl-1, 3-dioxolane | |
| 2-(bromomethyl)-1,3-dioxolane |
Specifications
| 4360-63-8 | |
| 175°C | |
| C4H7BrO2 | |
| MFCD00003214 | |
| 103309 | |
| 2-bromomethyl-1,3-dioxolane, bromoacetaldehyde ethylene acetal, 1,3-dioxolane, 2-bromomethyl, 1,3-dioxolan-2-yl methyl bromide, bromomethyl dioxolane, acmc-1afrf, 2-bromomethyl-1,3-dioxolan, ksc495q7b, 2-bromomethyl-1,3 dioxolane, 2-bromomethyl-1, 3-dioxolane | |
| CKIIJIDEWWXQEA-UHFFFAOYSA-N | |
| 2-(bromomethyl)-1,3-dioxolane | |
| 78068 | |
| 97% |
| 1.628 | |
| 62°C (145°F) | |
| 1.482 | |
| 25 g | |
| Moisture sensitive | |
| Immiscible with water. | |
| C1COC(O1)CBr | |
| 167.002 | |
| 167.01 | |
| 2-Bromomethyl-1,3-dioxolane |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H335
EINECSNumber : 224-443-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only