Learn More
2-Bromomethyl-1,3-dioxolane, 97%
CAS: 4360-63-8 | C4H7BrO2 | 167.002 g/mol
Supplier: Thermo Scientific Chemicals A1931222
Description
2-Bromomethyl-1,3-dioxolane is used in the preparation of alfa, beta-unsaturated aldehyde and 1, 4-two aldehyde monoacetals synthesis of monomers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Bromomethyl-1,3-dioxolane is used in the preparation of alfa, beta-unsaturated aldehyde and 1, 4-two aldehyde monoacetals synthesis of monomers.
Solubility
Immiscible with water.
Notes
Moisture sensitive. Incompatible with strong oxidizing agents, strong acids and strong bases.
Specifications
| 2-Bromomethyl-1,3-dioxolane | |
| 1.628 | |
| 62°C (145°F) | |
| 1.482 | |
| 100 g | |
| Moisture sensitive | |
| Immiscible with water. | |
| C1COC(O1)CBr | |
| 167.002 | |
| 167.01 |
| 4360-63-8 | |
| 175°C | |
| C4H7BrO2 | |
| MFCD00003214 | |
| 103309 | |
| 2-bromomethyl-1,3-dioxolane, bromoacetaldehyde ethylene acetal, 1,3-dioxolane, 2-bromomethyl, 1,3-dioxolan-2-yl methyl bromide, bromomethyl dioxolane, acmc-1afrf, 2-bromomethyl-1,3-dioxolan, ksc495q7b, 2-bromomethyl-1,3 dioxolane, 2-bromomethyl-1, 3-dioxolane | |
| CKIIJIDEWWXQEA-UHFFFAOYSA-N | |
| 2-(bromomethyl)-1,3-dioxolane | |
| 78068 | |
| 97% |
Chemical Identifiers
| 4360-63-8 | |
| 167.002 | |
| CKIIJIDEWWXQEA-UHFFFAOYSA-N | |
| 78068 | |
| C1COC(O1)CBr |
| C4H7BrO2 | |
| MFCD00003214 | |
| 2-bromomethyl-1,3-dioxolane, bromoacetaldehyde ethylene acetal, 1,3-dioxolane, 2-bromomethyl, 1,3-dioxolan-2-yl methyl bromide, bromomethyl dioxolane, acmc-1afrf, 2-bromomethyl-1,3-dioxolan, ksc495q7b, 2-bromomethyl-1,3 dioxolane, 2-bromomethyl-1, 3-dioxolane | |
| 2-(bromomethyl)-1,3-dioxolane |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H335
EINECSNumber : 224-443-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only