accessibility menu, dialog, popup

UserName

BrOO

CAS RN 4360-63-8

IUPAC Name: 2-(bromomethyl)-1,3-dioxolane
Synonyms: 2-(bromomethyl)-1,3-dioxolane
Molecular Weight (g/mol): 167
Molecular Formula: C4H7BrO2
InChi Key: CKIIJIDEWWXQEA-UHFFFAOYSA-N
SMILES: BrCC1OCCO1
Boiling Point 
  • (4)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (1)
  • (5)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (5)
  • (4)
  • (2)
  • (2)
  • (4)
  • (1)
  • (5)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)