missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(Bromomethyl)-1,3-benzothiazole, 95%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC06008DA
Specifications
| 2-(Bromomethyl)-1, 3-benzothiazole | |
| 95% | |
| C8H6BrNS | |
| 2-bromomethyl-1,3-benzothiazole, 2-bromomethyl benzo d thiazole, 2-bromomethyl benzothiazole, benzothiazole, 2-bromomethyl, benzo d thiazol-2-yl methyl bromide, zlchem 58, pubchem3912, acmc-1bpju, 2-bromomethylbenzothiazole | |
| C1=CC=C2C(=C1)N=C(S2)CBr | |
| 228.107 | |
| 228.11 |
| 106086-78-6 | |
| Amber Glass Bottle | |
| 1 g | |
| WFLCAOGKZQTOIG-UHFFFAOYSA-N | |
| 2-(bromomethyl)-1,3-benzothiazole | |
| 2776258 | |
| 95% |
Chemical Identifiers
| 106086-78-6 | |
| 228.107 | |
| 2-bromomethyl-1,3-benzothiazole, 2-bromomethyl benzo d thiazole, 2-bromomethyl benzothiazole, benzothiazole, 2-bromomethyl, benzo d thiazol-2-yl methyl bromide, zlchem 58, pubchem3912, acmc-1bpju, 2-bromomethylbenzothiazole | |
| 2-(bromomethyl)-1,3-benzothiazole |
| C8H6BrNS | |
| WFLCAOGKZQTOIG-UHFFFAOYSA-N | |
| 2776258 | |
| C1=CC=C2C(=C1)N=C(S2)CBr |