missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Bromoisonicotinic acid, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC437380050
Specifications
2-Bromoisonicotinic acid | |
Glass bottle | |
212.0°C to 222.0°C | |
66572-56-3 | |
100.0 | |
C6H4BrNO2 | |
YBTKGKVQEXAYEM-UHFFFAOYSA-N | |
2-bromopyridine-4-carboxylic acid | |
222701 | |
Crystals or Crystalline Powder |
Authentic | |
White | |
5g | |
96.0 | |
96% min. (HPLC) | |
2-bromisonicotins, 2-bromo-4-pyridine carboxylic acid, 2-bromo-4-pyridinecarboxylic acid, 2-bromo-isonicotinic acid, 2-bromoisonicotic acid, 2-bromoisonicotinic acid, 2-bromopyridine-4-carboxylicacid, 4-pyridinecarboxylic acid, 2-bromo, acmc-209nvt, pubchem9211 | |
C1=CN=C(C=C1C(=O)O)Br | |
202.01 | |
202.01 | |
97% |
Chemical Identifiers
66572-56-3 | |
202.01 | |
2-bromisonicotins, 2-bromo-4-pyridine carboxylic acid, 2-bromo-4-pyridinecarboxylic acid, 2-bromo-isonicotinic acid, 2-bromoisonicotic acid, 2-bromoisonicotinic acid, 2-bromopyridine-4-carboxylicacid, 4-pyridinecarboxylic acid, 2-bromo, acmc-209nvt, pubchem9211 | |
2-bromopyridine-4-carboxylic acid |
C6H4BrNO2 | |
YBTKGKVQEXAYEM-UHFFFAOYSA-N | |
222701 | |
C1=CN=C(C=C1C(=O)O)Br |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
GHS Signal Word: Warning