missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chemical Identifiers
| CAS | 66572-56-3 |
|---|---|
| Molecular Formula | C6H4BrNO2 |
| Molecular Weight (g/mol) | 202.01 |
| InChI Key | YBTKGKVQEXAYEM-UHFFFAOYSA-N |
| Synonym | 2-bromoisonicotinic acid, 2-bromo-4-pyridinecarboxylic acid, 2-bromo-isonicotinic acid, 2-bromo-4-pyridine carboxylic acid, 4-pyridinecarboxylic acid, 2-bromo, 2-bromoisonicotic acid, 2-bromisonicotins, 2-bromopyridine-4-carboxylicacid, pubchem9211, acmc-209nvt |
| PubChem CID | 222701 |
| IUPAC Name | 2-bromopyridine-4-carboxylic acid |
| SMILES | C1=CN=C(C=C1C(=O)O)Br |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
AC437380050
|
N/A | 5g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 66572-56-3 | |
| 202.01 | |
| 2-bromoisonicotinic acid, 2-bromo-4-pyridinecarboxylic acid, 2-bromo-isonicotinic acid, 2-bromo-4-pyridine carboxylic acid, 4-pyridinecarboxylic acid, 2-bromo, 2-bromoisonicotic acid, 2-bromisonicotins, 2-bromopyridine-4-carboxylicacid, pubchem9211, acmc-209nvt | |
| 2-bromopyridine-4-carboxylic acid |
| C6H4BrNO2 | |
| YBTKGKVQEXAYEM-UHFFFAOYSA-N | |
| 222701 | |
| C1=CN=C(C=C1C(=O)O)Br |
Specifications
| 66572-56-3 | |
| 100.0 | |
| 96% min. (HPLC) | |
| 2-bromoisonicotinic acid, 2-bromo-4-pyridinecarboxylic acid, 2-bromo-isonicotinic acid, 2-bromo-4-pyridine carboxylic acid, 4-pyridinecarboxylic acid, 2-bromo, 2-bromoisonicotic acid, 2-bromisonicotins, 2-bromopyridine-4-carboxylicacid, pubchem9211, acmc-209nvt | |
| C1=CN=C(C=C1C(=O)O)Br | |
| 202.01 | |
| 202.01 | |
| 2-Bromoisonicotinic acid |
| 96.0 | |
| Authentic | |
| C6H4BrNO2 | |
| YBTKGKVQEXAYEM-UHFFFAOYSA-N | |
| 2-bromopyridine-4-carboxylic acid | |
| 222701 | |
| 97% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
GHS Signal Word: Warning
RUO – Research Use Only