accessibility menu, dialog, popup

UserName

HOONBr

CAS RN 66572-56-3

IUPAC Name: 2-bromopyridine-4-carboxylic acid
Synonyms: 2-bromopyridine-4-carboxylic acid
Molecular Weight (g/mol): 202.01
Molecular Formula: C6H4BrNO2
InChi Key: YBTKGKVQEXAYEM-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=NC(Br)=C1
Molecular Weight (g/mol) 
  • (4)
  • (1)
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (2)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (4)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)