missing translation for 'onlineSavingsMsg'
Learn More

(2-Bromoethyl)triphenylphosphonium bromide, 98%

Catalog No. AAB2483314
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2483314 25 g
AAB2483306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2483314 Supplier Thermo Scientific Chemicals Supplier No. B2483314
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7301-93-1 | C20H19Br2P | 450.15 g/mol

(2-Bromoethyl)triphenylphosphonium bromide is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(2-Bromoethyl)triphenylphosphonium bromide is used as pharmaceutical intermediates.

Notes
Hygroscopic. Store away from moisture and oxidizing agents under dry inert gas. Protect from humidity and water. Incompatible with oxidizing agents and moisture.

Chemical Identifiers

CAS 7301-93-1
Molecular Formula C20H19Br2P
Molecular Weight (g/mol) 450.15
MDL Number MFCD04039352
InChI Key ZHLVWYZWQPNQDQ-UHFFFAOYSA-M
Synonym 2-bromoethyl triphenylphosphonium bromide, phosphonium, 2-bromoethyl triphenyl-, bromide, 2-bromoethyl triphenylphosohonium bromide, 2-bromoethyl triphenylphosphanium bromide, 2-bromoethyl triphenyl phosphanium bromide, 2-bromoethyl triphenyl phosphonium bromide, phosphonium, 2-bromoethyl triphenyl-,bromide
PubChem CID 22210408
IUPAC Name 2-bromoethyl(triphenyl)phosphanium;bromide
SMILES [Br-].BrCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 186°C to 189°C
Quantity 25 g
Sensitivity Hygroscopic
Formula Weight 450.15
Percent Purity 98%
Chemical Name or Material (2-Bromoethyl)triphenylphosphonium bromide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only