missing translation for 'onlineSavingsMsg'
Learn More

2-Bromobutane 98.0+%, TCI America™

Catalog No. p-7118527
Change view
Click to view available options
Quantity:
25 g
500 g
Catalog No. Quantity
B056125G 25 g
B0561500G 500 g
2 options

Catalog No. B056125G

Supplier: TCI America B056125G

Only null left

Chemical Identifiers

CAS 78-76-2
Molecular Formula C4H9Br
Molecular Weight (g/mol) 137.02
MDL Number MFCD00000156
InChI Key UPSXAPQYNGXVBF-UHFFFAOYSA-N
Synonym sec-butyl bromide, butane, 2-bromo, 2-butyl bromide, methylethylbromomethane, 2-bromo-butane, secondary butyl bromide, sec-butylbromide, 1-bromo-1-methylpropane, ccris 106, bromobutane, 2
PubChem CID 6554
IUPAC Name 2-bromobutane
SMILES CCC(C)Br

Specifications

Color Yellow
Boiling Point 91°C
Quantity 25 g
UN Number 2339
Formula Weight 137.02
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromobutane
EINECSNumber (2)-0074
RTECSNumber EJ6228000
TSCA Yes