accessibility menu, dialog, popup

UserName

H3CCH3Br

CAS RN 78-76-2

IUPAC Name: 2-bromobutane
Synonyms: 2-bromobutane
Molecular Weight (g/mol): 137.02
Molecular Formula: C4H9Br
InChi Key: UPSXAPQYNGXVBF-UHFFFAOYNA-N
SMILES: CCC(C)Br
Boiling Point 
  • (5)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (5)
  • (5)
  • (2)
  • (2)
  • (2)
  • (3)
  • (7)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)