missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromobenzylamine Hydrochloride 98.0+%, TCI America™
$119.30 - $404.51
Chemical Identifiers
| CAS | 5465-63-4 |
|---|---|
| Molecular Formula | C7H8BrClN |
| Molecular Weight (g/mol) | 221.50 |
| MDL Number | MFCD00012853 |
| InChI Key | ZANQNQMPTMEAIP-UHFFFAOYSA-N |
| Synonym | 2-bromobenzylamine hydrochloride, 2-bromophenyl methanamine hydrochloride, o-bromobenzylamine hydrochloride, 1-2-bromophenyl methanamine hydrochloride, 2-bromobenzylamine hcl, 1-2-bromophenyl methylamine hydrochloride, 2-bromophenyl methylamine, chloride, benzenemethanamine, 2-bromo-, hydrochloride, pubchem3749, acmc-1aup2 |
| PubChem CID | 2724202 |
| IUPAC Name | 1-(2-bromophenyl)methanamine hydrochloridyl |
| SMILES | [Cl].NCC1=CC=CC=C1Br |
Chemical Identifiers
| 5465-63-4 | |
| 221.50 | |
| ZANQNQMPTMEAIP-UHFFFAOYSA-N | |
| 2724202 | |
| [Cl].NCC1=CC=CC=C1Br |
| C7H8BrClN | |
| MFCD00012853 | |
| 2-bromobenzylamine hydrochloride, 2-bromophenyl methanamine hydrochloride, o-bromobenzylamine hydrochloride, 1-2-bromophenyl methanamine hydrochloride, 2-bromobenzylamine hcl, 1-2-bromophenyl methylamine hydrochloride, 2-bromophenyl methylamine, chloride, benzenemethanamine, 2-bromo-, hydrochloride, pubchem3749, acmc-1aup2 | |
| 1-(2-bromophenyl)methanamine hydrochloridyl |
Specifications
| 5465-63-4 | |
| C7H8BrClN | |
| 5 g | |
| ZANQNQMPTMEAIP-UHFFFAOYSA-N | |
| 1-(2-bromophenyl)methanamine hydrochloridyl | |
| 2724202 | |
| ≥98.0% (T) | |
| 2-Bromobenzylamine Hydrochloride |
| White-Yellow | |
| MFCD00012853 | |
| 2-bromobenzylamine hydrochloride, 2-bromophenyl methanamine hydrochloride, o-bromobenzylamine hydrochloride, 1-2-bromophenyl methanamine hydrochloride, 2-bromobenzylamine hcl, 1-2-bromophenyl methylamine hydrochloride, 2-bromophenyl methylamine, chloride, benzenemethanamine, 2-bromo-, hydrochloride, pubchem3749, acmc-1aup2 | |
| [Cl].NCC1=CC=CC=C1Br | |
| 221.50 | |
| 222.51 | |
| Crystalline Powder |
Safety and Handling
TSCA : No