accessibility menu, dialog, popup

UserName

ClH2NBr

CAS RN 5465-63-4

IUPAC Name: 1-(2-bromophenyl)methanamine hydrochloridyl
Synonyms: 1-(2-bromophenyl)methanamine hydrochloridyl
Molecular Weight (g/mol): 221.5
Molecular Formula: C7H8BrClN
InChi Key: ZANQNQMPTMEAIP-UHFFFAOYSA-N
SMILES: [Cl].NCC1=CC=CC=C1Br
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)