missing translation for 'onlineSavingsMsg'
Learn More

2-Bromobenzylamine Hydrochloride 98.0+%, TCI America™

Catalog No. B18365G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B18365G 5 g
B183625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B18365G Supplier TCI America Supplier No. B18365G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5465-63-4
Molecular Formula C7H8BrClN
Molecular Weight (g/mol) 221.50
MDL Number MFCD00012853
InChI Key ZANQNQMPTMEAIP-UHFFFAOYSA-N
Synonym 2-bromobenzylamine hydrochloride, 2-bromophenyl methanamine hydrochloride, o-bromobenzylamine hydrochloride, 1-2-bromophenyl methanamine hydrochloride, 2-bromobenzylamine hcl, 1-2-bromophenyl methylamine hydrochloride, 2-bromophenyl methylamine, chloride, benzenemethanamine, 2-bromo-, hydrochloride, pubchem3749, acmc-1aup2
PubChem CID 2724202
IUPAC Name 1-(2-bromophenyl)methanamine hydrochloridyl
SMILES [Cl].NCC1=CC=CC=C1Br

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 222.51
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromobenzylamine Hydrochloride