Learn More
2-Bromobenzylamine hydrochloride, 97%
CAS: 5465-63-4 | C7H8BrClN | 221.50 g/mol
$141.59 - $471.34
Chemical Identifiers
| CAS | 5465-63-4 |
|---|---|
| Molecular Formula | C7H8BrClN |
| Molecular Weight (g/mol) | 221.50 |
| MDL Number | MFCD00012853 |
| InChI Key | ZANQNQMPTMEAIP-UHFFFAOYSA-N |
| Synonym | 2-bromobenzylamine hydrochloride, 2-bromophenyl methanamine hydrochloride, o-bromobenzylamine hydrochloride, 1-2-bromophenyl methanamine hydrochloride, 2-bromobenzylamine hcl, 1-2-bromophenyl methylamine hydrochloride, 2-bromophenyl methylamine, chloride, benzenemethanamine, 2-bromo-, hydrochloride, pubchem3749, acmc-1aup2 |
| PubChem CID | 2724202 |
| SMILES | [Cl].NCC1=CC=CC=C1Br |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1815106
|
Thermo Scientific Chemicals
A1815106 |
5 g |
Each for $141.59
|
|
|||||
|
AAA1815114
|
Thermo Scientific Chemicals
A1815114 |
25 g |
Each for $471.34
|
|
|||||
Description
It is employed as an intermediate in organic syntheses.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is employed as an intermediate in organic syntheses.
Solubility
Soluble in methanol (25 mg/ml).
Notes
Hygroscopic. Store away from oxidizing agents, water/moisture. Store under inert gas. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
| 5465-63-4 | |
| 221.50 | |
| ZANQNQMPTMEAIP-UHFFFAOYSA-N | |
| 2724202 |
| C7H8BrClN | |
| MFCD00012853 | |
| 2-bromobenzylamine hydrochloride, 2-bromophenyl methanamine hydrochloride, o-bromobenzylamine hydrochloride, 1-2-bromophenyl methanamine hydrochloride, 2-bromobenzylamine hcl, 1-2-bromophenyl methylamine hydrochloride, 2-bromophenyl methylamine, chloride, benzenemethanamine, 2-bromo-, hydrochloride, pubchem3749, acmc-1aup2 | |
| [Cl].NCC1=CC=CC=C1Br |
Specifications
| 5465-63-4 | |
| C7H8BrClN | |
| 5 g | |
| Hygroscopic | |
| Soluble in methanol (25mg/ml). | |
| [Cl].NCC1=CC=CC=C1Br | |
| 221.50 | |
| 222.52 | |
| 2-Bromobenzylamine hydrochloride |
| 227°C to 230°C | |
| MFCD00012853 | |
| 3913456 | |
| 2-bromobenzylamine hydrochloride, 2-bromophenyl methanamine hydrochloride, o-bromobenzylamine hydrochloride, 1-2-bromophenyl methanamine hydrochloride, 2-bromobenzylamine hcl, 1-2-bromophenyl methylamine hydrochloride, 2-bromophenyl methylamine, chloride, benzenemethanamine, 2-bromo-, hydrochloride, pubchem3749, acmc-1aup2 | |
| ZANQNQMPTMEAIP-UHFFFAOYSA-N | |
| (2-bromophenyl)methanamine;hydrochloride | |
| 2724202 | |
| 97% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only