missing translation for 'onlineSavingsMsg'
Learn More

2-Bromobenzylamine hydrochloride, 97%

Catalog No. AAA1815106
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1815106 5 g
AAA1815114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1815106 Supplier Thermo Scientific Chemicals Supplier No. A1815106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 5465-63-4 | C7H8BrClN | 221.50 g/mol

It is employed as an intermediate in organic syntheses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as an intermediate in organic syntheses.

Solubility
Soluble in methanol (25 mg/ml).

Notes
Hygroscopic. Store away from oxidizing agents, water/moisture. Store under inert gas. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 5465-63-4
Molecular Formula C7H8BrClN
Molecular Weight (g/mol) 221.50
MDL Number MFCD00012853
InChI Key ZANQNQMPTMEAIP-UHFFFAOYSA-N
Synonym 2-bromobenzylamine hydrochloride, 2-bromophenyl methanamine hydrochloride, o-bromobenzylamine hydrochloride, 1-2-bromophenyl methanamine hydrochloride, 2-bromobenzylamine hcl, 1-2-bromophenyl methylamine hydrochloride, 2-bromophenyl methylamine, chloride, benzenemethanamine, 2-bromo-, hydrochloride, pubchem3749, acmc-1aup2
PubChem CID 2724202
SMILES [Cl].NCC1=CC=CC=C1Br

Specifications

Melting Point 227°C to 230°C
Quantity 5 g
Beilstein 3913456
Sensitivity Hygroscopic
Solubility Information Soluble in methanol (25mg/ml).
IUPAC Name (2-bromophenyl)methanamine;hydrochloride
Formula Weight 222.52
Percent Purity 97%
Chemical Name or Material 2-Bromobenzylamine hydrochloride
Hazard Category H302+H312-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only