missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromobenzyl Mercaptan 98.0+%, TCI America™
$213.69 - $643.00
Chemical Identifiers
| CAS | 143888-85-1 |
|---|---|
| Molecular Formula | C7H7BrS |
| Molecular Weight (g/mol) | 203.10 |
| MDL Number | MFCD04038925 |
| InChI Key | UJNSDLRPHRMVGZ-UHFFFAOYSA-N |
| Synonym | 2-bromobenzyl mercaptan, 2-bromobenzylmercaptan, 2-bromophenyl methanethiol, o-bromobenzyl hydrosulfide, 2-bromobenzylthiol, 2-bromophenyl methane-1-thiol, 2-bromobenzyl, 2-bromobenzyl mercap, pubchem6847 |
| PubChem CID | 2773673 |
| IUPAC Name | (2-bromophenyl)methanethiol |
| SMILES | SCC1=CC=CC=C1Br |
Chemical Identifiers
| 143888-85-1 | |
| 203.10 | |
| UJNSDLRPHRMVGZ-UHFFFAOYSA-N | |
| 2773673 | |
| SCC1=CC=CC=C1Br |
| C7H7BrS | |
| MFCD04038925 | |
| 2-bromobenzyl mercaptan, 2-bromobenzylmercaptan, 2-bromophenyl methanethiol, o-bromobenzyl hydrosulfide, 2-bromobenzylthiol, 2-bromophenyl methane-1-thiol, 2-bromobenzyl, 2-bromobenzyl mercap, pubchem6847 | |
| (2-bromophenyl)methanethiol |
Specifications
| 143888-85-1 | |
| 82°C | |
| MFCD04038925 | |
| 2810 | |
| UJNSDLRPHRMVGZ-UHFFFAOYSA-N | |
| (2-bromophenyl)methanethiol | |
| 2773673 | |
| ≥98.0% (GC) | |
| 2-Bromobenzyl Mercaptan |
| Yellow | |
| C7H7BrS | |
| 5 g | |
| 2-bromobenzyl mercaptan, 2-bromobenzylmercaptan, 2-bromophenyl methanethiol, o-bromobenzyl hydrosulfide, 2-bromobenzylthiol, 2-bromophenyl methane-1-thiol, 2-bromobenzyl, 2-bromobenzyl mercap, pubchem6847 | |
| SCC1=CC=CC=C1Br | |
| 203.10 | |
| 203.10 | |
| Liquid |
Safety and Handling
TSCA : No