missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromobenzophenone 97.0+%, TCI America™
$39.14 - $152.86
Chemical Identifiers
| CAS | 13047-06-8 |
|---|---|
| Molecular Formula | C13H9BrO |
| Molecular Weight (g/mol) | 261.12 |
| MDL Number | MFCD00032462 |
| InChI Key | ABEVIHIQUUXDMS-UHFFFAOYSA-N |
| PubChem CID | 2735534 |
| IUPAC Name | (2-bromophenyl)(phenyl)methanone |
| SMILES | BrC1=CC=CC=C1C(=O)C1=CC=CC=C1 |
Chemical Identifiers
| 13047-06-8 | |
| 261.12 | |
| ABEVIHIQUUXDMS-UHFFFAOYSA-N | |
| (2-bromophenyl)(phenyl)methanone |
| C13H9BrO | |
| MFCD00032462 | |
| 2735534 | |
| BrC1=CC=CC=C1C(=O)C1=CC=CC=C1 |
Specifications
| 13047-06-8 | |
| Yellow | |
| C13H9BrO | |
| 1 g | |
| BrC1=CC=CC=C1C(=O)C1=CC=CC=C1 | |
| 261.12 | |
| 261.12 | |
| Crystalline Lumps |
| 42°C | |
| 163°C | |
| MFCD00032462 | |
| ABEVIHIQUUXDMS-UHFFFAOYSA-N | |
| (2-bromophenyl)(phenyl)methanone | |
| 2735534 | |
| ≥97.0% (GC) | |
| 2-Bromobenzophenone |
Safety and Handling
TSCA : No