accessibility menu, dialog, popup

UserName

CAS RN 13047-06-8

BrO

CAS RN 13047-06-8

IUPAC Name: (2-bromophenyl)(phenyl)methanone
Synonyms: (2-bromophenyl)(phenyl)methanone
Molecular Weight (g/mol): 261.12
Molecular Formula: C13H9BrO
InChi Key: ABEVIHIQUUXDMS-UHFFFAOYSA-N
SMILES: BrC1=CC=CC=C1C(=O)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results