Learn More
2-Bromobenzaldehyde, 98%
CAS: 6630-33-7 | C7H5BrO | 185.02 g/mol
Supplier: Thermo Scientific Chemicals A1506522
Description
2-Bromobenzaldehyde is used in L-threonine aldolase-catalyzed enantio and diastereoselective aldol reactions. Further, it reacts with trichloromethane to prepare 1-(2-bromo-phenyl)-2,2,2-trichloro-ethanol.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Bromobenzaldehyde is used in L-threonine aldolase-catalyzed enantio and diastereoselective aldol reactions. Further, it reacts with trichloromethane to prepare 1-(2-bromo-phenyl)-2,2,2-trichloro-ethanol.
Solubility
Soluble in alcohol and benzene. Insoluble in water.
Notes
Air sensitive. Incompatible with strong bases, strong oxidizing agents and strong reducing agents.
Specifications
| 2-Bromobenzaldehyde | |
| 18°C to 21°C | |
| 228°C to 230°C | |
| Characteristic | |
| 1.596 | |
| 100 g | |
| Air Sensitive | |
| Soluble in alcohol and benzene. Insoluble in water. | |
| BrC1=CC=CC=C1C=O | |
| 185.02 | |
| 185.03 |
| 6630-33-7 | |
| 1.597 | |
| 95°C (203°F) | |
| C7H5BrO | |
| MFCD00003300 | |
| 636132 | |
| o-bromobenzaldehyde, benzaldehyde, 2-bromo, benzaldehyde, o-bromo, 2-bromo benzaldehyde, bromobenzaldehyde, bromo benzaldehyde, o-bromo-benzaldehyde, 2-formylbromobenzene, 2-bromo-benzaldehyde, benzaldehyde, bromo | |
| NDOPHXWIAZIXPR-UHFFFAOYSA-N | |
| 2-bromobenzaldehyde | |
| 81129 | |
| 98% |
Chemical Identifiers
| 6630-33-7 | |
| 185.02 | |
| NDOPHXWIAZIXPR-UHFFFAOYSA-N | |
| 81129 | |
| BrC1=CC=CC=C1C=O |
| C7H5BrO | |
| MFCD00003300 | |
| o-bromobenzaldehyde, benzaldehyde, 2-bromo, benzaldehyde, o-bromo, 2-bromo benzaldehyde, bromobenzaldehyde, bromo benzaldehyde, o-bromo-benzaldehyde, 2-formylbromobenzene, 2-bromo-benzaldehyde, benzaldehyde, bromo | |
| 2-bromobenzaldehyde |
Safety and Handling
GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 229-622-2
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only