missing translation for 'onlineSavingsMsg'
Learn More

2-Bromobenzaldehyde, 98%

Catalog No. p-7022340
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1506514 25 g
AAA1506522 100 g
AAA1506536 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1506514 Supplier Thermo Scientific Chemicals Supplier No. A1506514
Only null left

CAS: 6630-33-7 | C7H5BrO | 185.02 g/mol

2-Bromobenzaldehyde is used in L-threonine aldolase-catalyzed enantio and diastereoselective aldol reactions. Further, it reacts with trichloromethane to prepare 1-(2-bromo-phenyl)-2,2,2-trichloro-ethanol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromobenzaldehyde is used in L-threonine aldolase-catalyzed enantio and diastereoselective aldol reactions. Further, it reacts with trichloromethane to prepare 1-(2-bromo-phenyl)-2,2,2-trichloro-ethanol.

Solubility
Soluble in alcohol and benzene. Insoluble in water.

Notes
Air sensitive. Incompatible with strong bases, strong oxidizing agents and strong reducing agents.

Chemical Identifiers

CAS 6630-33-7
Molecular Formula C7H5BrO
Molecular Weight (g/mol) 185.02
MDL Number MFCD00003300
InChI Key NDOPHXWIAZIXPR-UHFFFAOYSA-N
Synonym o-bromobenzaldehyde, benzaldehyde, 2-bromo, benzaldehyde, o-bromo, 2-bromo benzaldehyde, bromobenzaldehyde, bromo benzaldehyde, o-bromo-benzaldehyde, 2-formylbromobenzene, 2-bromo-benzaldehyde, benzaldehyde, bromo
PubChem CID 81129
IUPAC Name 2-bromobenzaldehyde
SMILES BrC1=CC=CC=C1C=O

Specifications

Melting Point 18°C to 21°C
Density 1.597
Boiling Point 228°C to 230°C
Flash Point 95°C (203°F)
Odor Characteristic
Refractive Index 1.596
Quantity 25 g
Beilstein 636132
Sensitivity Air Sensitive
Solubility Information Soluble in alcohol and benzene. Insoluble in water.
Formula Weight 185.03
Percent Purity 98%
Chemical Name or Material 2-Bromobenzaldehyde
Show More Show Less
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 229-622-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only