missing translation for 'onlineSavingsMsg'
Learn More

2-Bromoanthracene 97.0+%, TCI America™

Catalog No. p-7119695
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B26161G 1 g
B26165G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B26161G Supplier TCI America Supplier No. B26161G

Chemical Identifiers

CAS 7321-27-9
Molecular Formula C14H9Br
Molecular Weight (g/mol) 257.13
MDL Number MFCD07784002
InChI Key PYXBCVWIECUMDW-UHFFFAOYSA-N
Synonym 2-bromo-anthracene, anthracene, 2-bromo, pubchem19987, acmc-209opw, ksc377a5l
PubChem CID 12346099
IUPAC Name 2-bromoanthracene
SMILES BrC1=CC2=CC3=CC=CC=C3C=C2C=C1

Specifications

Melting Point 220°C
Quantity 1 g
Formula Weight 257.13
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromoanthracene