accessibility menu, dialog, popup

UserName

CAS RN 7321-27-9

IUPAC Name: 2-bromoanthracene
Synonyms: 2-bromoanthracene
Molecular Weight (g/mol): 257.13
Molecular Formula: C14H9Br
InChi Key: PYXBCVWIECUMDW-UHFFFAOYSA-N
SMILES: BrC1=CC2=CC3=CC=CC=C3C=C2C=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)