missing translation for 'onlineSavingsMsg'
Learn More

2-Bromoaniline 98.0+%, TCI America™

Catalog No. B054125G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B054125G 25 g
B0541250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B054125G Supplier TCI America Supplier No. B054125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 615-36-1
Molecular Formula C6H6BrN
Molecular Weight (g/mol) 172.025
MDL Number MFCD00007632
InChI Key AOPBDRUWRLBSDB-UHFFFAOYSA-N
Synonym o-bromoaniline, benzenamine, 2-bromo, o-aminobromobenzene, aniline, o-bromo, 2-bromobenzenamine, bromoaniline 2-, 2-bromo aniline, 2-bromophenylamine, benzenamine,2-bromo, 2-bromioaniline
PubChem CID 11992
IUPAC Name 2-bromoaniline
SMILES C1=CC=C(C(=C1)N)Br

Specifications

Color White-Yellow
Boiling Point 229°C
Quantity 25 g
UN Number 2811
Formula Weight 172.03
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 2-Bromoaniline
EINECSNumber (3)-0221
TSCA Yes