accessibility menu, dialog, popup

UserName

NH2Br

CAS RN 615-36-1

IUPAC Name: 2-bromoaniline
Synonyms: O-bromoaniline
Molecular Weight (g/mol): 172.03
Molecular Formula: C6H6BrN
InChi Key: AOPBDRUWRLBSDB-UHFFFAOYSA-N
SMILES: NC1=CC=CC=C1Br
Boiling Point 
  • (3)
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (5)
Percent Purity 
  • (2)
  • (5)
  • (1)
Quantity 
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (3)
  • (2)
  • (3)
  • (3)
  • (5)
  • (2)
  • (5)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)