missing translation for 'onlineSavingsMsg'
Learn More

2-Bromoaniline, 98%

Catalog No. p-4229096
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC106610250 25 g Glass bottle
AC106611000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC106610250 Supplier Thermo Scientific Chemicals Supplier No. 106610250
Only null left

CAS: 615-36-1 | C6H6BrN | 172.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 615-36-1
Molecular Formula C6H6BrN
Molecular Weight (g/mol) 172.02
InChI Key AOPBDRUWRLBSDB-UHFFFAOYSA-N
PubChem CID 11992
IUPAC Name 2-bromoaniline
SMILES C1=CC=C(C(=C1)N)Br

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 29.0°C to 31.0°C
Density 1.5780g/mL
Boiling Point 229.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.6170 to 1.6190
Linear Formula BrC6H4NH2
Quantity 25 g
Beilstein 12,631
Specific Gravity 1.578
Synonym o-bromoaniline, benzenamine, 2-bromo, o-aminobromobenzene, aniline, o-bromo, 2-bromobenzenamine, bromoaniline 2-, 2-bromo aniline, 2-bromophenylamine, benzenamine,2-bromo, 2-bromioaniline
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ether,alcohols
Formula Weight 172.02
Percent Purity 98%
Physical Form Liquid After Melting
Chemical Name or Material 2-Bromoaniline
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse
EINECSNumber 210-421-3

RUO – Research Use Only