missing translation for 'onlineSavingsMsg'
Learn More

2-Bromoadamantane 97.0+%, TCI America™

$350.38 - $350.38

Chemical Identifiers

CAS 7314-85-4
Molecular Formula C10H15Br
Molecular Weight (g/mol) 215.134
MDL Number MFCD00074745
InChI Key RCXJARRRXOPXBC-UHFFFAOYSA-N
Synonym 2-adamantyl bromide, adamantane, 2-bromo, tricyclo 3.3.1.13,7 decane, 2-bromo, tricyclo 3.3.1.1 3,7 decane, 2-bromo, 1r,3r,5r,7r-2-bromoadamantane, 2-bromo-adamantane, 2-bromanyladamantane, pubchem8752, 2alpha-bromoadamantane, 2-bromotricyclo 3.3.1.13.7 decane
PubChem CID 522482
IUPAC Name 2-bromoadamantane
SMILES C1C2CC3CC1CC(C2)C3Br
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B03805G
View Documents
TCI America
B03805G
5 g
Each for $350.38
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7314-85-4
215.134
RCXJARRRXOPXBC-UHFFFAOYSA-N
522482
C1C2CC3CC1CC(C2)C3Br
C10H15Br
MFCD00074745
2-adamantyl bromide, adamantane, 2-bromo, tricyclo 3.3.1.13,7 decane, 2-bromo, tricyclo 3.3.1.1 3,7 decane, 2-bromo, 1r,3r,5r,7r-2-bromoadamantane, 2-bromo-adamantane, 2-bromanyladamantane, pubchem8752, 2alpha-bromoadamantane, 2-bromotricyclo 3.3.1.13.7 decane
2-bromoadamantane

Specifications

7314-85-4
White-Yellow
MFCD00074745
2-adamantyl bromide, adamantane, 2-bromo, tricyclo 3.3.1.13,7 decane, 2-bromo, tricyclo 3.3.1.1 3,7 decane, 2-bromo, 1r,3r,5r,7r-2-bromoadamantane, 2-bromo-adamantane, 2-bromanyladamantane, pubchem8752, 2alpha-bromoadamantane, 2-bromotricyclo 3.3.1.13.7 decane
C1C2CC3CC1CC(C2)C3Br
215.134
215.13
Crystalline Powder
140°C
C10H15Br
5 g
RCXJARRRXOPXBC-UHFFFAOYSA-N
2-bromoadamantane
522482
≥97.0% (GC)
2-Bromoadamantane
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'