missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromoadamantane 97.0+%, TCI America™
Supplier: TCI America B03805G
Specifications
| 2-Bromoadamantane | |
| 140°C | |
| C10H15Br | |
| 5 g | |
| RCXJARRRXOPXBC-UHFFFAOYSA-N | |
| 2-bromoadamantane | |
| 522482 | |
| ≥97.0% (GC) |
| 7314-85-4 | |
| White-Yellow | |
| MFCD00074745 | |
| 2-adamantyl bromide, adamantane, 2-bromo, tricyclo 3.3.1.13,7 decane, 2-bromo, tricyclo 3.3.1.1 3,7 decane, 2-bromo, 1r,3r,5r,7r-2-bromoadamantane, 2-bromo-adamantane, 2-bromanyladamantane, pubchem8752, 2alpha-bromoadamantane, 2-bromotricyclo 3.3.1.13.7 decane | |
| C1C2CC3CC1CC(C2)C3Br | |
| 215.134 | |
| 215.13 | |
| Crystalline Powder |
Chemical Identifiers
| 7314-85-4 | |
| 215.134 | |
| RCXJARRRXOPXBC-UHFFFAOYSA-N | |
| 522482 | |
| C1C2CC3CC1CC(C2)C3Br |
| C10H15Br | |
| MFCD00074745 | |
| 2-adamantyl bromide, adamantane, 2-bromo, tricyclo 3.3.1.13,7 decane, 2-bromo, tricyclo 3.3.1.1 3,7 decane, 2-bromo, 1r,3r,5r,7r-2-bromoadamantane, 2-bromo-adamantane, 2-bromanyladamantane, pubchem8752, 2alpha-bromoadamantane, 2-bromotricyclo 3.3.1.13.7 decane | |
| 2-bromoadamantane |
Safety and Handling
TSCA : No