missing translation for 'onlineSavingsMsg'
Learn More

2-Bromoacrylic acid, 95%

CAS: 10443-65-9 | C3H3BrO2 | 150.96 g/mol

$129.22 - $584.32

Chemical Identifiers

CAS 10443-65-9
Molecular Formula C3H3BrO2
Molecular Weight (g/mol) 150.96
MDL Number MFCD00014333
InChI Key HMENQNSSJFLQOP-UHFFFAOYSA-N
Synonym 2-bromoacrylic acid, bromoacrylic acid, ccris 5478, .alpha.-bromoacrylic acid, 2-bromo-acrylic acid, alpha-bromo acrylic acid, 2-propenoic acid, bromo, acmc-1c0k4, 2-propenoic acid, 2-bromo
PubChem CID 82633
IUPAC Name 2-bromoprop-2-enoic acid
SMILES OC(=O)C(Br)=C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0119406
View Documents
Thermo Scientific Chemicals
L0119406
5 g
Each for $129.22
Only null left
 
AAL0119414
View Documents
Thermo Scientific Chemicals
L0119414
25 g
Each for $584.32
Only null left
 
Description

Description

2-Bromoacrylic acid is used as a raw material on QSAR models to predict mutagenicity of acrylates. It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromoacrylic acid is used as a raw material on QSAR models to predict mutagenicity of acrylates. It is used as a pharmaceutical intermediate.

Solubility
It is miscible with water.

Notes
Heat sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
Specifications

Chemical Identifiers

10443-65-9
150.96
HMENQNSSJFLQOP-UHFFFAOYSA-N
82633
OC(=O)C(Br)=C
C3H3BrO2
MFCD00014333
2-bromoacrylic acid, bromoacrylic acid, ccris 5478, .alpha.-bromoacrylic acid, 2-bromo-acrylic acid, alpha-bromo acrylic acid, 2-propenoic acid, bromo, acmc-1c0k4, 2-propenoic acid, 2-bromo
2-bromoprop-2-enoic acid

Specifications

10443-65-9
C3H3BrO2
5 g
1740582
It is miscible with water.
OC(=O)C(Br)=C
150.96
150.96
2-Bromoacrylic acid
65°C to 70°C
MFCD00014333
UN3261
2-bromoacrylic acid, bromoacrylic acid, ccris 5478, .alpha.-bromoacrylic acid, 2-bromo-acrylic acid, alpha-bromo acrylic acid, 2-propenoic acid, bromo, acmc-1c0k4, 2-propenoic acid, 2-bromo
HMENQNSSJFLQOP-UHFFFAOYSA-N
2-bromoprop-2-enoic acid
82633
95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H317
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.

P260-P264b-P271-P272-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P333+P313-P363-P501c

H314-H317-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 233-928-1

TSCA : No

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only