CAS RN 10443-65-9
IUPAC Name:
2-bromoprop-2-enoic acid
Synonyms:
2-bromoprop-2-enoic acid
Molecular Weight (g/mol):
150.96
Molecular Formula:
C3H3BrO2
InChi Key:
HMENQNSSJFLQOP-UHFFFAOYSA-N
SMILES:
OC(=O)C(Br)=C
1
–
1
of
1
results
1
Promotions Available
2-Bromoacrylic acid, 95%
2-Bromoacrylic acid, 95%
CAS: 10443-65-9 Molecular Formula: C3H3BrO2 Molecular Weight (g/mol): 150.96 MDL Number: MFCD00014333 InChI Key: HMENQNSSJFLQOP-UHFFFAOYSA-N Synonym: 2-bromoacrylic acid,bromoacrylic acid,ccris 5478,.alpha.-bromoacrylic acid,2-bromo-acrylic acid,alpha-bromo acrylic acid,2-propenoic acid, bromo,acmc-1c0k4,2-propenoic acid, 2-bromo PubChem CID: 82633 IUPAC Name: 2-bromoprop-2-enoic acid SMILES: OC(=O)C(Br)=C