missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-9,9-dimethylfluorene 98.0+%, TCI America™
Supplier: TCI America B38945G
Specifications
| 2-Bromo-9,9-dimethylfluorene | |
| 69°C | |
| 190°C | |
| MFCD09261272 | |
| 2-bromo-9,9-dimethyl-9h-fluorene, 9,9-dimethyl-2-bromofluorene, 9h-fluorene, 2-bromo-9,9-dimethyl, 2-bromo-9,9-dimethyl-fluorene, 2-bromo-9,9-dimethylfluoren, ksc494q3r, 2-bromo-9,9-dimethyl fluorene, 9,9-dimethyl-2-bromofluorenone, 2-bromo-9,9-dimethylfluorene | |
| CC1(C2=CC=CC=C2C3=C1C=C(C=C3)Br)C | |
| 273.173 | |
| 273.17 | |
| Crystalline Powder |
| 28320-31-2 | |
| White-Yellow | |
| C15H13Br | |
| 5 g | |
| MBHPOBSZPYEADG-UHFFFAOYSA-N | |
| 2-bromo-9,9-dimethylfluorene | |
| 10945606 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 28320-31-2 | |
| 273.173 | |
| MBHPOBSZPYEADG-UHFFFAOYSA-N | |
| 10945606 | |
| CC1(C2=CC=CC=C2C3=C1C=C(C=C3)Br)C |
| C15H13Br | |
| MFCD09261272 | |
| 2-bromo-9,9-dimethyl-9h-fluorene, 9,9-dimethyl-2-bromofluorene, 9h-fluorene, 2-bromo-9,9-dimethyl, 2-bromo-9,9-dimethyl-fluorene, 2-bromo-9,9-dimethylfluoren, ksc494q3r, 2-bromo-9,9-dimethyl fluorene, 9,9-dimethyl-2-bromofluorenone, 2-bromo-9,9-dimethylfluorene | |
| 2-bromo-9,9-dimethylfluorene |
Safety and Handling
TSCA : No