missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-9,9-dimethylfluorene 98.0+%, TCI America™

Catalog No. p-7123230
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B38945G 5 g
B389425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B38945G Supplier TCI America Supplier No. B38945G
Only null left

Chemical Identifiers

CAS 28320-31-2
Molecular Formula C15H13Br
Molecular Weight (g/mol) 273.173
MDL Number MFCD09261272
InChI Key MBHPOBSZPYEADG-UHFFFAOYSA-N
Synonym 2-bromo-9,9-dimethyl-9h-fluorene, 9,9-dimethyl-2-bromofluorene, 9h-fluorene, 2-bromo-9,9-dimethyl, 2-bromo-9,9-dimethyl-fluorene, 2-bromo-9,9-dimethylfluoren, ksc494q3r, 2-bromo-9,9-dimethyl fluorene, 9,9-dimethyl-2-bromofluorenone, 2-bromo-9,9-dimethylfluorene
PubChem CID 10945606
IUPAC Name 2-bromo-9,9-dimethylfluorene
SMILES CC1(C2=CC=CC=C2C3=C1C=C(C=C3)Br)C

Specifications

Melting Point 69°C
Color White-Yellow
Boiling Point 190°C
Quantity 5 g
Formula Weight 273.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-9,9-dimethylfluorene