missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-9,9-dihexylfluorene 97.0+%, TCI America™
$98.79 - $288.94
Chemical Identifiers
| CAS | 226070-05-9 |
|---|---|
| Molecular Formula | C25H33Br |
| Molecular Weight (g/mol) | 413.443 |
| MDL Number | MFCD14584649 |
| InChI Key | NNYSRQDAPSNOKV-UHFFFAOYSA-N |
| PubChem CID | 18674600 |
| IUPAC Name | 2-bromo-9,9-dihexylfluorene |
| SMILES | CCCCCCC1(C2=CC=CC=C2C3=C1C=C(C=C3)Br)CCCCCC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
B4404200MG
|
TCI America
B4404200MG |
200 mg |
Each for $98.79
|
|
|||||
|
B44041G
|
TCI America
B44041G |
1 g |
Each for $288.94
|
|
|||||
Chemical Identifiers
| 226070-05-9 | |
| 413.443 | |
| NNYSRQDAPSNOKV-UHFFFAOYSA-N | |
| 2-bromo-9,9-dihexylfluorene |
| C25H33Br | |
| MFCD14584649 | |
| 18674600 | |
| CCCCCCC1(C2=CC=CC=C2C3=C1C=C(C=C3)Br)CCCCCC |
Specifications
| 226070-05-9 | |
| MFCD14584649 | |
| NNYSRQDAPSNOKV-UHFFFAOYSA-N | |
| 2-bromo-9,9-dihexylfluorene | |
| 18674600 | |
| ≥97.0% (GC) | |
| 2-Bromo-9,9-dihexylfluorene |
| C25H33Br | |
| 200 mg | |
| CCCCCCC1(C2=CC=CC=C2C3=C1C=C(C=C3)Br)CCCCCC | |
| 413.443 | |
| 413.44 | |
| Liquid |
Safety and Handling
TSCA : No