accessibility menu, dialog, popup

UserName

CH3BrCH3

CAS RN 226070-05-9

IUPAC Name: 2-bromo-9,9-dihexyl-9H-fluorene
Synonyms: 2-bromo-9,9-dihexylfluorene
Molecular Weight (g/mol): 413.44
Molecular Formula: C25H33Br
InChi Key: NNYSRQDAPSNOKV-UHFFFAOYSA-N
SMILES: CCCCCCC1(CCCCCC)C2=CC=CC=C2C2=C1C=C(Br)C=C2
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results