missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-9,9-dihexylfluorene 97.0+%, TCI America™

Catalog No. B4404200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4404200MG 200 mg
B44041G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B4404200MG Supplier TCI America Supplier No. B4404200MG
Only null left

Chemical Identifiers

CAS 226070-05-9
Molecular Formula C25H33Br
Molecular Weight (g/mol) 413.443
MDL Number MFCD14584649
InChI Key NNYSRQDAPSNOKV-UHFFFAOYSA-N
PubChem CID 18674600
IUPAC Name 2-bromo-9,9-dihexylfluorene
SMILES CCCCCCC1(C2=CC=CC=C2C3=C1C=C(C=C3)Br)CCCCCC

Specifications

Quantity 200 mg
Formula Weight 413.44
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromo-9,9-dihexylfluorene