missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-6-methylbenzoic acid, 98%

CAS: 90259-31-7 | C8H7BrO2 | 215.05 g/mol

Supplier:  Thermo Scientific Chemicals L2014906

 View more versions of this product

Catalog No. AAL2014906


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Bromo-6-methylbenzoic acid
107°C to 111°C
MFCD01310788
2614094
ICXBPDJQFPIBSS-UHFFFAOYSA-N
2-bromo-6-methylbenzoic acid
2735589
98%
90259-31-7
C8H7BrO2
5 g
6-bromo-o-toluic acid, 2-bromo-6-methyl-benzoic acid, 6-bromo-2-methylbenzoic acid, benzoic acid, 2-bromo-6-methyl, bromtoluylsaure, bromotoluic acid, pubchem4715, acmc-209r5e, ksc494k9r, 3-bromo-2-carboxytoluene
CC1=C(C(O)=O)C(Br)=CC=C1
215.05
215.05

Chemical Identifiers

90259-31-7
215.05
ICXBPDJQFPIBSS-UHFFFAOYSA-N
2735589
CC1=C(C(O)=O)C(Br)=CC=C1
C8H7BrO2
MFCD01310788
6-bromo-o-toluic acid, 2-bromo-6-methyl-benzoic acid, 6-bromo-2-methylbenzoic acid, benzoic acid, 2-bromo-6-methyl, bromtoluylsaure, bromotoluic acid, pubchem4715, acmc-209r5e, ksc494k9r, 3-bromo-2-carboxytoluene
2-bromo-6-methylbenzoic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only