missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-6-methylbenzoic acid, 98%

Catalog No. p-7048041
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL2014903 1 g
AAL2014906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL2014903 Supplier Thermo Scientific Chemicals Supplier No. L2014903
Only null left

CAS: 90259-31-7 | C8H7BrO2 | 215.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 90259-31-7
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.05
MDL Number MFCD01310788
InChI Key ICXBPDJQFPIBSS-UHFFFAOYSA-N
Synonym 6-bromo-o-toluic acid, 2-bromo-6-methyl-benzoic acid, 6-bromo-2-methylbenzoic acid, benzoic acid, 2-bromo-6-methyl, bromtoluylsaure, bromotoluic acid, pubchem4715, acmc-209r5e, ksc494k9r, 3-bromo-2-carboxytoluene
PubChem CID 2735589
IUPAC Name 2-bromo-6-methylbenzoic acid
SMILES CC1=C(C(O)=O)C(Br)=CC=C1

Specifications

Melting Point 107°C to 111°C
Quantity 1 g
Beilstein 2614094
Formula Weight 215.05
Percent Purity 98%
Chemical Name or Material 2-Bromo-6-methylbenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only