missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-6-methoxybenzoic acid, 98%

Catalog No. AAH3160403
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3160403 1 g
AAH3160406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3160403 Supplier Thermo Scientific Chemicals Supplier No. H3160403
Only null left

CAS: 31786-45-5 | C8H7BrO3 | 231.045 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 31786-45-5
Molecular Formula C8H7BrO3
Molecular Weight (g/mol) 231.045
MDL Number MFCD00800732
InChI Key BNYFDICJYCBPRW-UHFFFAOYSA-N
Synonym 2-bromo-6-methoxybenzoicacid, benzoic acid, 2-bromo-6-methoxy, pubchem11265, acmc-209hoz, ksc494q5t
PubChem CID 11117881
IUPAC Name 2-bromo-6-methoxybenzoic acid
SMILES COC1=C(C(=CC=C1)Br)C(=O)O

Specifications

Quantity 1 g
Formula Weight 231.05
Percent Purity 98%
Chemical Name or Material 2-Bromo-6-methoxybenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only