CAS RN 31786-45-5
IUPAC Name:
2-bromo-6-methoxybenzoic acid
Synonyms:
2-bromo-6-methoxybenzoic acid
Molecular Weight (g/mol):
231.05
Molecular Formula:
C8H7BrO3
InChi Key:
BNYFDICJYCBPRW-UHFFFAOYSA-N
SMILES:
COC1=C(C(O)=O)C(Br)=CC=C1
1
–
1
of
1
results
1
Promotions Available
2-Bromo-6-methoxybenzoic acid, 98%
2-Bromo-6-methoxybenzoic acid, 98%
CAS: 31786-45-5 Molecular Formula: C8H7BrO3 Molecular Weight (g/mol): 231.045 MDL Number: MFCD00800732 InChI Key: BNYFDICJYCBPRW-UHFFFAOYSA-N Synonym: 2-bromo-6-methoxybenzoicacid,benzoic acid, 2-bromo-6-methoxy,pubchem11265,acmc-209hoz,ksc494q5t PubChem CID: 11117881 IUPAC Name: 2-bromo-6-methoxybenzoic acid SMILES: COC1=C(C(=CC=C1)Br)C(=O)O