missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-6-fluorobenzotrifluoride 97.0+%, TCI America™
$54.06 - $156.59
Chemical Identifiers
| CAS | 261951-85-3 |
|---|---|
| Molecular Formula | C7H3BrF4 |
| Molecular Weight (g/mol) | 242.999 |
| MDL Number | MFCD01631569 |
| InChI Key | LOXSSZUHCPBDMD-UHFFFAOYSA-N |
| Synonym | 2-bromo-6-fluorobenzotrifluoride, 1-bromo-3-fluoro-2-trifluoromethyl benzene, 3-fluoro-2-trifluoromethyl bromobenzene, benzene, 1-bromo-3-fluoro-2-trifluoromethyl, 2-bromo-alpha,alpha,alpha,6-tetrafluorotoluene, 1-bromo-3-fluoro-2-trifluoromethyl-benzene, pubchem1638, 2-bromo-6-fluorobenzo trifluoride |
| PubChem CID | 2773345 |
| IUPAC Name | 1-bromo-3-fluoro-2-(trifluoromethyl)benzene |
| SMILES | C1=CC(=C(C(=C1)Br)C(F)(F)F)F |
Chemical Identifiers
| 261951-85-3 | |
| 242.999 | |
| LOXSSZUHCPBDMD-UHFFFAOYSA-N | |
| 2773345 | |
| C1=CC(=C(C(=C1)Br)C(F)(F)F)F |
| C7H3BrF4 | |
| MFCD01631569 | |
| 2-bromo-6-fluorobenzotrifluoride, 1-bromo-3-fluoro-2-trifluoromethyl benzene, 3-fluoro-2-trifluoromethyl bromobenzene, benzene, 1-bromo-3-fluoro-2-trifluoromethyl, 2-bromo-alpha,alpha,alpha,6-tetrafluorotoluene, 1-bromo-3-fluoro-2-trifluoromethyl-benzene, pubchem1638, 2-bromo-6-fluorobenzo trifluoride | |
| 1-bromo-3-fluoro-2-(trifluoromethyl)benzene |
Specifications
| 261951-85-3 | |
| C7H3BrF4 | |
| 1 g | |
| LOXSSZUHCPBDMD-UHFFFAOYSA-N | |
| 1-bromo-3-fluoro-2-(trifluoromethyl)benzene | |
| 2773345 | |
| ≥97.0% (GC) | |
| 2-Bromo-6-fluorobenzotrifluoride |
| Yellow | |
| MFCD01631569 | |
| 2-bromo-6-fluorobenzotrifluoride, 1-bromo-3-fluoro-2-trifluoromethyl benzene, 3-fluoro-2-trifluoromethyl bromobenzene, benzene, 1-bromo-3-fluoro-2-trifluoromethyl, 2-bromo-alpha,alpha,alpha,6-tetrafluorotoluene, 1-bromo-3-fluoro-2-trifluoromethyl-benzene, pubchem1638, 2-bromo-6-fluorobenzo trifluoride | |
| C1=CC(=C(C(=C1)Br)C(F)(F)F)F | |
| 242.999 | |
| 243.00 | |
| Liquid |
Safety and Handling
TSCA : No